ContactUsLinkComponent
VWR® Signature™, Disposable electroporation cuvettes
VWR® Signature™, Disposable electroporation cuvettes
Catalog # VWRU89047-208
Supplier:  VWR Collection
undefined
VWR® Signature™, Disposable electroporation cuvettes
Catalog # VWRU89047-208
Supplier:  VWR Collection
Specifications
  • 容量:
    400 µl
  • 説明:
    Electroporation cuvettes with 2 mm gap
  • Pk:
    50
Specifications
About this item
High quality durable polycarbonate has been tested to optimise transformation efficiencies. These cuvettes are built to provide exact tolerances ensuring reproducible results.
  • Polished aluminum plates reducing static and arcing
  • Compatible with most electroporation devices
  • Gamma irradiated to warranty sterility
Packaging: 50 individually wrapped cuvettes.