Orders LinkContactUsLinkComponent
スライドホルダ、VWR®製
Supplier:   VWR Collection
スライドホルダ、VWR®製
Supplier:  VWR Collection
Certificates
スライドホルダ、VWR®製
スライドホルダ、VWR®製
2 Options Available Below
Product Details & Documents
Polypropylene resistant slide holders easy to open for durable protection.
Certifications: FDA certified
Packaging: 250 slides per box, four boxes per case.
Product Family Options
Product Information
  • 説明
    Colour
    Material
    No. of slides
    Availability
    Price
  • Single slide holders
    Blue
    PP
    1
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items - $89.28
    $89.28
    1 * 250 items
       $89.28
  • Double slide holders
    Clear
    PP
    2
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items - $104.36
    $104.36
    1 * 250 items
       $104.36