ロッドエンドコネクター、Talon®、VWR®、ROS-ID™
VWRU60079-010
- For:13 mm Ø rods
- 説明:Rod end connector
- Pk:1
This nickel-plated zinc connector holds rods firmly at 90°.
- Use this rod end connector when semi-permanent installations are required
- Comes with two set screws and is precision bored for close fit with 13 mm (¹/₂") rods