スパチュラ、VWR
178 mm
Catalog # VWRU82027-534
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stainless steel, PTFE coated. This spatula features one rounded end and one flat end, with PTFE coating throughout.
Can be used to transfer corrosive or sticky materials without scratching or damaging container surfaces.
Specifications
- 長さ:178 mm