スクープ、セパレートハンドル付き、VWR®
Stainless steel.
- Sturdy stainless steel construction
- Handle and scoop are sold separately
The rounded tip of this scoop makes it easy to push into powders or break up solidified chemicals into smaller pieces, while its concave shape makes it ideal for picking them up.
Stainless steel.
- Sturdy stainless steel construction
- Handle and scoop are sold separately
The rounded tip of this scoop makes it easy to push into powders or break up solidified chemicals into smaller pieces, while its concave shape makes it ideal for picking them up.
Product Family Options
- 説明長さ
Handle
- 説明長さ