Orders LinkContactUsLinkComponent
オーバーオール、VWRシグネチャー
オーバーオール、VWRシグネチャー
オーバーオール、VWRシグネチャー
オーバーオール、VWRシグネチャー
These protective, durable, and comfortable overalls made with DuPont™ Tyvek® IsoClean® material provide an excellent barrier against dry particulates, non hazardous liquids, and microorganisms. Garments are chemically and biologically inert, low-linting, and static-free until laundered.
  • Excellent dry particulate, nonhazardous liquid, and microorganism barrier
  • Chemically and biologically inert
  • Low-linting, static-free
  • Serged seams, laydown collar, front zipper closure, and elastic wrists
  • Available with elastic ancles or attached Gripper™ boots
  • Boot attachments combine an embossed PVC exterior sole with a coarsely textured interior to ensure a secure, non slip fit and feel
  • Catalog No:
  • VWRU89012-822
  • VWRU89012-824
  • VWRU89012-826
  • VWRU89012-828
  • VWRU89012-830
  • VWRU80200-624
  • VWRU89012-832
  • VWRU80200-626
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • Garment application:
  • Laboratory|Industrial
  • Laboratory|Industrial
  • Laboratory|Industrial
  • Laboratory|Industrial
  • Laboratory|Industrial
  • Laboratory|Industrial
  • Laboratory|Industrial
  • Laboratory|Industrial
Product Family Options
  • 説明
    Colour
    衣服のサイズ
  • Overalls with elastic ankles
    White
    S
    1 * 25 items
    1 * 25 items
    Specifications
    Cat. No.
    VWRU89012-822
    Anti-static
    +
    Garment application
    Laboratory|Industrial
  • Overalls with elastic ankles
    White
    M
    1 * 25 items
    1 * 25 items
    Specifications
    Cat. No.
    VWRU89012-824
    Anti-static
    +
    Garment application
    Laboratory|Industrial
  • Overalls with elastic ankles
    White
    L
    1 * 25 items
    1 * 25 items
    Specifications
    Cat. No.
    VWRU89012-826
    Anti-static
    +
    Garment application
    Laboratory|Industrial
  • Overalls with elastic ankles
    White
    XL
    1 * 25 items
    1 * 25 items
    Specifications
    Cat. No.
    VWRU89012-828
    Anti-static
    +
    Garment application
    Laboratory|Industrial
  • Overalls with elastic ankles
    White
    XXL
    1 * 25 items
    1 * 25 items
    Specifications
    Cat. No.
    VWRU89012-830
    Anti-static
    +
    Garment application
    Laboratory|Industrial
  • Overalls with attached boots
    White
    XL
    1 * 25 items
    1 * 25 items
    Specifications
    Cat. No.
    VWRU80200-624
    Anti-static
    +
    Garment application
    Laboratory|Industrial
  • Overalls with elastic ankles
    White
    3XL
    1 * 25 items
    1 * 25 items - $594.85
    $594.85
    1 * 25 items
       $594.85
    Specifications
    Cat. No.
    VWRU89012-832
    Anti-static
    +
    Garment application
    Laboratory|Industrial
  • Overalls with attached boots
    White
    XXL
    1 * 25 items
    1 * 25 items
    Specifications
    Cat. No.
    VWRU80200-626
    Anti-static
    +
    Garment application
    Laboratory|Industrial