秤量ボート、VWR® (英語)
Supplier: VWR Collection
Jump to:
About this item
Thin-walled glass.
- Very good chemical resistance
- Good temperature resistance
- Flattened base provides stable rest on balance pan
Ideal for speedy, precise weighing and transfer of samples without loss.
2 Options Available Below
Product Details & Documents
Thin-walled glass.
- Very good chemical resistance
- Good temperature resistance
- Flattened base provides stable rest on balance pan
Ideal for speedy, precise weighing and transfer of samples without loss.
Product Family Options
Product Information
- 容量長さ外径AvailabilityPrice