ストッパー、ゴム製、1穴付き
5,0 mm, 54,0 mm, 64,0 mm
Catalog # VWRU59584-354
Supplier: VWR Collection
Jump to:
Product Details & Documents
These stoppers contain less free sulphur and have a lower density than black rubber stoppers. Its composition ensures high resilency and holes facilitates the laboratory usage. Ideal for food grade applications such as beer and wine making used with carboys.
- Natural gum rubber
- White
Specifications
- Ø bottom:54,0 mm
- Ø top:64,0 mm
- 穴の直径:5,0 mm