Orders LinkContactUsLinkComponent
ピペット用バルブ, VWR® (英語)
ピペット用バルブ, VWR® (英語)
Supplier:   VWR Collection
ピペット用バルブ, VWR® (英語)
Supplier:  VWR Collection
Certificates
ピペット用バルブ, VWR® (英語)
ピペット用バルブ, VWR® (英語)
2 Options Available Below
Product Details & Documents
Pipette bulbs made of natural rubber.
  • Durable material
  • Bulbs are suitable for medicine droppers and small dropping pipettes
  • Neck is grooved to take flat flange of pipettes
  • Heavy-walled, cylindrical in shape
  • Bulbs fit pipettes 6 to 8 mm Ext.Ø
Product Family Options
Product Information
  • 説明
    Colour
    Availability
    Price
  • Dropper bulbs, 1 ml
    Black
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items
  • Dropper bulbs, 2 ml
    Red
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items