Orders LinkContactUsLinkComponent
VWR®, ビーカー、ローフォーム、PMP
VWR®, ビーカー、ローフォーム、PMP
VWR®, ビーカー、ローフォーム、PMP
VWR®, ビーカー、ローフォーム、PMP
PMP, crystal clear, graduated.
  • Excellent chemical resistance
  • Autoclavable
  • Withstand a temperature of up to 170 °C for short periods
With large spout and easy to read blue graduations.
 : Conform to ISO 7056 1981 (E) and BS 5404 Part 1.
Product Family Options
  • 容量
    目盛り間隔
  • 250 ml
    10 ml
    1 * 6 items
    1 * 6 items - $154.46
    $154.46
    1 * 6 items
       $154.46
  • 500 ml
    10 ml
    1 * 3 items
    1 * 3 items - $138.99
    $138.99
    1 * 3 items
       $138.99
  • 1000 ml
    20 ml
    1 * 3 items
    1 * 3 items - $214.95
    $214.95
    1 * 3 items
       $214.95
  • 5000 ml
    500 ml
    1 * 1 items
    1 * 1 items - $190.52
    $190.52
    1 * 1 items
       $190.52