微量遠沈管、1.5mL
1,5 ml, Orange, with attached lid, graduated, conical bottom, non sterile
VWRU89000-034
PP, disposable, graduated.
- RNase- and DNase-free
- Autoclavable
- Tubes can be centrifuged to 20000×g
Tubes feature flat caps and frosted areas on the sides for easy labelling. Snap caps can be penetrated by a syringe. Easy-to-open caps will not pop open during centrifuging.
: Manufactured in a Class 100000 cleanroom environment. Manufactured under ISO 13485:2016 and ISO 9001:2015 quality management systems. Meet the requirements of RoHS.
: All tubes packaged in resealable bags.
- Type:with attached lid
- Version:graduated, conical bottom, non sterile
- 容量:1,5 ml
- Colour:Orange
- Seq:06
- Cat. No.:VWRU89000-034
- Environmentally Preferable:Low Manufacturing Impact|Sustainable Packaging