マグネット式スターラーバー、Spinvane® (スピンベーン)
PTFE covered, Alnico V magnetic core. Spinvane® Triangle shaped magnetic stirring bars are designed for stirring in test tubes, centrifuge tubes and micro vials.
- The wings can also be trimmed to size if needed
- Autoclave safe
PTFE covered, Alnico V magnetic core. Spinvane® Triangle shaped magnetic stirring bars are designed for stirring in test tubes, centrifuge tubes and micro vials.
- The wings can also be trimmed to size if needed
- Autoclave safe
Product Family Options
- For