Orders LinkContactUsLinkComponent
VWR®, Specimen forceps, serrated tips
VWR®, Specimen forceps, serrated tips
Supplier:   VWR Collection
VWR®, Specimen forceps, serrated tips
Supplier:  VWR Collection
Certificates
VWR®, Specimen forceps, serrated tips
VWR®, Specimen forceps, serrated tips
7 Options Available Below
Product Details & Documents
These specimen forceps made from stainless steel are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
  • Serrated tips
  • Easy to use
Product Family Options
Product Information
  • Type
    長さ
    Availability
    Price
  • Straight, serrated tips
    114 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $6.84
    $6.84
    1 * 1 items
       $6.84
  • Straight, serrated tips
    152 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $19.54
    $19.54
    1 * 1 items
       $19.54
  • Straight, serrated tips
    178 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $15.23
    $15.23
    1 * 1 items
       $15.23
  • Straight, serrated tips
    203 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $31.26
    $31.26
    1 * 1 items
       $31.26
  • Curved, serrated tips
    203 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $20.01
    $20.01
    1 * 1 items
       $20.01
  • Straight, serrated tips
    254 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $24.94
    $24.94
    1 * 1 items
       $24.94
  • Straight, serrated tips
    305 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $28.71
    $28.71
    1 * 1 items
       $28.71