クランプ、3本爪、Talon® II
Supplier: VWR Collection
Certificates
About this item
Steel, nickel plated. These clamps holds irregular shapes easily. They feature precise pressure regulation to reduce glassware breakage.
- Open yoke design allows full depth of the clamp to be used, unobstructed by the yoke, springs, or adjusting screws
- Narrow profile on single adjustment clamps; variable holding angle on dual adjustment clamps
- Design is easy to clean and decontaminate
- Distinctive nickel finish for maximum protection against corrosion
Delivery: Supplied with non slip vinyl sleeves and fibreglass covers for temperatures above 100 °C.
Frequently Bought Together