Orders LinkContactUsLinkComponent
クランプ、2本爪、VWR®製
クランプ、2本爪、VWR®製
Supplier:   VWR Collection
クランプ、2本爪、VWR®製
Supplier:  VWR Collection
Certificates
クランプ、2本爪、VWR®製
クランプ、2本爪、VWR®製
5 Options Available Below
Product Details & Documents
Zinc, nickel plated. These clamps with single or dual adjustment securely holds laboratory glassware and apparatus. Ideal for holding beakers, flasks, and test tubes.
  • Large grip adjustment range
  • Two-prong construction
Extension arm attaches clamp head securely and offers easy positioning in the deepest fume hoods. Clamps are constructed with round extension arms that allow the clamps to be rotated 360°. Extension arms also allow placement of apparatus at various distances from lab frames without compromising the integrity of the experiment.
Delivery information: Supplied with non slip vinyl sleeves, and for temperatures above 100 °C (212 °F), fiberglass covers. An additional holder must be purchased in order to attach clamps to frames or other apparatus.
Product Family Options
Single adjustment
Product Information
  • 説明
    Spanning width
    長さ
    Availability
    Price
  • Single adjustment clamps
    0 - 92
    229 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $69.41
    $69.41
    1 * 1 items
       $69.41
Dual adjustment
Product Information
  • 説明
    Spanning width
    長さ
    Availability
    Price
  • Dual adjustment clamps
    0 - 75 mm
    229 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $63.34
    $63.34
    1 * 1 items
       $63.34
  • Dual adjustment clamps
    0 - 95 mm
    248 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $69.41
    $69.41
    1 * 1 items
       $69.41
  • Dual adjustment clamps
    0 - 75 mm
    229 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $267.22
    $267.22
    1 * 1 items
       $267.22
  • Dual adjustment clamps
    0 - 95 mm
    248 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $321.56
    $321.56
    1 * 1 items
       $321.56