Orders LinkContactUsLinkComponent
微量遠沈管、1.7/2.0 ml
微量遠沈管、1.7/2.0 ml
Supplier:   VWR Collection
微量遠沈管、1.7/2.0 ml
Supplier:  VWR Collection
Certificates
微量遠沈管、1.7/2.0 ml
2 Options Available Below
Product Details & Documents
PP, graduated, autoclavable and freezable with attached flat caps.
  • DNase-, RNase and endotoxin-free
  • Can withstand centrifugation to RCF 20000×g
  • Fit all standard and high capacity rotors
  • Tubes with X-Resin™ provides ultra-low binding characteristics
Medical grade superior clarity for visualisation of samples and isolated pellets. Graduated frosted side panel and flat snap cap provide surface for labelling.
Product Family Options
Product Information
  • 容量
    Colour
    Type
    Version
    Sterile
    Availability
    Price
  • 1,7 ml
    Colourless
    with attached lid
    graduated, conical bottom, non sterile
    -
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $33.87
    $33.87
    1 * 500 items
       $33.87
  • 2,0 ml
    Colourless
    with attached lid
    graduated, conical bottom, non sterile
    -
    Log in for availability
    Log in for availability
    1 * 400 items
    1 * 400 items - $33.87
    $33.87
    1 * 400 items
       $33.87