マグネット式ラベルロール、Write-On
Write-On/wash-off labels, Vinyl, White, 15,2 m×25 mm
Catalog # VWRU80094-278
Supplier: VWR Collection
Jump to:
Product Details & Documents
This magnetic label material has a white vinyl surface that may be written on with wet-erase marker and then wiped off. Provided in a roll that may be cut to any size strip.
- Thickness: 0,8 mm
Specifications
- Colour:White
- 説明:Write-On/wash-off labels
- 寸法:15,2 m×25 mm
- Material:Vinyl