Orders LinkContactUsLinkComponent
解剖用鉗子、VWR®製
Supplier:   VWR Collection
解剖用鉗子、VWR®製
Supplier:  VWR Collection
Certificates
解剖用鉗子、VWR®製
解剖用鉗子、VWR®製
9 Options Available Below
Product Details & Documents
Dissecting forceps made of stainless steel. Available straight or curved and in different lengths.
Product Family Options
Product Information
  • Type
    長さ
    Availability
    Price
  • Curved, fine tips
    114 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $19.67
    $19.67
    1 * 1 items
       $19.67
  • Straight, fine tips
    114 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $6.27
    $6.27
    1 * 1 items
       $6.27
  • Straight, serrated, medium tips (blunt)
    114 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $6.92
    $6.92
    1 * 1 items
       $6.92
  • Straight, round tips
    114 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $16.35
    $16.35
    1 * 1 items
       $16.35
  • Straight, serrated, medium pointed tips
    114 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $5.58
    $5.58
    1 * 1 items
       $5.58
  • Straight, serrated, with fine tips
    114 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Curved, serrated, with fine tips
    114 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Straight, round tips
    152 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $13.65
    $13.65
    1 * 1 items
       $13.65
  • Curved, serrated, fine tips
    152 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items