ビーカートング、 ステンレス鋼
318 mm
Catalog # VWRU82027-374
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stainless steel, fibreglass coating. Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 ml capacities.
Tongs feature a stainless steel construction, a riveted joint, and fibreglass coating for use in high-temperature applications.
Specifications
- 長さ:318 mm