Orders LinkContactUsLinkComponent
Büchner製ファンネル
Büchner製ファンネル
Supplier:   VWR Collection
Büchner製ファンネル
Supplier:  VWR Collection
Certificates
Büchner製ファンネル
Büchner製ファンネル
5 Options Available Below
Product Details & Documents
Porcelain, glased inside and out, except rim.
  • Resistant to chemicals
  • Very good temperature resistance
  • For paper or membrane filtration discs
Product Family Options
Product Information
  • 容量
    For filter Ø
    Availability
    Price
  • 87 ml
    50 - 55 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $64.56
    $64.56
    1 * 1 items
       $64.56
  • 186 ml
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $84.85
    $84.85
    1 * 1 items
       $84.85
  • 550 ml
    110 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items
  • 700 ml
    110 - 124 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $197.38
    $197.38
    1 * 1 items
       $197.38
  • 1860 ml
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $435.05
    $435.05
    1 * 1 items
       $435.05