流出対応キット、HazMat
Hazmat lab kit, 34,1 litres/kit
Catalog # VWRU89168-998
Supplier: VWR Collection
Jump to:
Product Details & Documents
This portable kit handles all types of lab spills.
Certifications: Supports compliance with 40 CFR 263.30(a) safety standard.
Delivery information: Kit contains twenty pads, three socks, one pair nitrile gloves, one pair safety goggles, and one water-resistant yellow bag with tie.
Specifications
- 説明:Hazmat lab kit
- Absorbency:34,1 litres/kit