Orders LinkContactUsLinkComponent
ポーラスボトムるつぼ、VWR®
Supplier:   VWR Collection
ポーラスボトムるつぼ、VWR®
Supplier:  VWR Collection
Certificates
ポーラスボトムるつぼ、VWR®
ポーラスボトムるつぼ、VWR®
2 Options Available Below
Product Details & Documents
These porcelain crucibles feature a porous bottom with a porous disc permanently formed into the walls. Designed with stable porosity and flow rate.
 : Lids need to be ordered seperately.
Product Family Options
Product Information
  • Type
    容量
    Ø×H
    細孔サイズ
    Availability
    Price
  • Porous bottom
    25 ml
    36×42 mm
    15 µm
    1 * 1 items
  • Porous bottom
    25 ml
    36×42 mm
    1,2 µm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $90.22
    $90.22
    1 * 1 items
       $90.22