Orders LinkContactUsLinkComponent
VWR®, バイオハザード標本袋
VWR®, バイオハザード標本袋
Supplier:   VWR Collection
VWR®, バイオハザード標本袋
Supplier:  VWR Collection
Certificates
VWR®, バイオハザード標本袋
VWR®, バイオハザード標本袋
2 Options Available Below
Product Details & Documents
Designed for safe and secure transport of a specimen container and documentation, these 51 µm thick bags feature a pouch to keep the paperwork separate from the container. They have a self-sealing closure and are tamper evident.
Product Family Options
Product Information
  • 説明
    Material
    長さ×幅
    厚さ
    Availability
    Price
  • Biohazard specimen bags
    LDPE
    229×152,4 mm
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $205.91
    $205.91
    1 * 500 items
       $205.91
  • Biohazard specimen bags
    LDPE
    305×229 mm
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $323.91
    $323.91
    1 * 500 items
       $323.91