Orders LinkContactUsLinkComponent
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
Supplier:   VWR Collection
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
Supplier:  VWR Collection
Certificates
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
VWR® Tri-Pour®, ビーカー、 使い捨て、 PP (トライポアー)
5 Options Available Below
Product Details & Documents
PP, graduated.
  • With three pouring lips for dripless dispensing
  • Graduatations with accuracy of ±5%
  • Write-on cardboard caps ensure spillproof, leak-resistant closure
These beakers may also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.
Product Family Options
Product Information
  • 容量
    Availability
    Price
  • 50 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $45.48
    $45.48
    1 * 100 items
       $45.48
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU25384-150
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $76.55
    $76.55
    1 * 100 items
       $76.55
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU25384-154
  • 400 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $110.38
    $110.38
    1 * 100 items
       $110.38
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU25384-156
  • 800 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $167.38
    $167.38
    1 * 100 items
       $167.38
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU25384-158
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $213.93
    $213.93
    1 * 100 items
       $213.93
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU25384-160