VWR®, Wash Bottles, Wide Neck, with Coloured Caps
Supplier: VWR Collection
Certificates
About this item
LDPE, with colour coded PP spray caps.
- Available with spray caps in five colours
- Colour-coded closures for easy identification
- Wide mouths for easy filling
Frequently Bought Together