Orders LinkContactUsLinkComponent
ボーメ比重計、デュラク™、Bcl-2 (ヒト)
ボーメ比重計、デュラク™、Bcl-2 (ヒト)
Supplier:   VWR Collection
ボーメ比重計、デュラク™、Bcl-2 (ヒト)
Supplier:  VWR Collection
Certificates
ボーメ比重計、デュラク™、Bcl-2 (ヒト)
ボーメ比重計、デュラク™、Bcl-2 (ヒト)
2 Options Available Below
Product Details & Documents
Baumé hydrometer designed for determining the specific gravity and Baumé in liquids lighter then water.
  • Dual scale
  • Individually serialised
  • Lead free metal ballast
Specially used in chemistry, pharmacology, food and beverage, and in the starch industry.
Certifications: NIST and DAkkS. Calibration conforms to A2LA accreditions.
納品情報: Supplied with a multi-language statement of accuracy.
Specifications for Product Family
Specifications
  • Catalog No:
  • VWRU34670-140
  • VWRU34670-161
  • Reference temperature:
  • 15,6 °C
Specifications
Product Family Options
Product Information
  • 説明
    Range
    Division
    長さ
    Availability
    Price
  • Baumé hydrometer, Durac™
    0,790 - 0,900 g/l ; 24 - 45 °Bé
    0,002 g/l ; 0,5 °Bé
    165 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $54.79
    $54.79
    1 * 1 items
       $54.79
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU34670-140
    Reference temperature
    15,6 °C
  • Baumé hydrometer, Durac™
    0,890 - 1,000 g/l ; 10 - 25 °Bé
    0,002 g/l ; 0,5 °Bé
    165 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $54.58
    $54.58
    1 * 1 items
       $54.58
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU34670-161
    Reference temperature