Orders LinkContactUsLinkComponent
ビーカー、低形状、PP
ビーカー、低形状、PP
Supplier:   VWR Collection
ビーカー、低形状、PP
Supplier:  VWR Collection
Certificates
ビーカー、低形状、PP
ビーカー、低形状、PP
7 Options Available Below
Product Details & Documents
PP, translucent, graduated.
  • Features no-drip spout for clean pouring
  • Tapered walls for secure grip
  • With an oval marking spot
Ideal for general laboratory procedures. These are easy-to-handle stackable beakers and is break-resistant, nontoxic, non-contaminating, and extremely chemical-resistant. Sturdy construction can withstand repeated autoclaving.
Product Family Options
Product Information
  • 容量
    目盛り範囲
    Availability
    Price
  • 100 ml
    10 - 100 ml
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - $75.29
    $75.29
    1 * 12 items
       $75.29
  • 150 ml
    20 - 150 ml
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - $97.17
    $97.17
    1 * 12 items
       $97.17
  • 250 ml
    25 - 250 ml
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $65.20
    $65.20
    1 * 6 items
       $65.20
  • 600 ml
    100 - 600 ml
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items - $83.24
    $83.24
    1 * 4 items
       $83.24
  • 1000 ml
    100 - 1000 ml
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items - $77.14
    $77.14
    1 * 3 items
       $77.14
  • 2000 ml
    200 - 2000 ml
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
  • 4000 ml
    500 - 4000 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $104.17
    $104.17
    1 * 1 items
       $104.17