Orders LinkContactUsLinkComponent
VWR®, マイクロチューブ収納ボックス、
VWR®, マイクロチューブ収納ボックス、
Supplier:  VWR Collection
Certificates
VWR®, マイクロチューブ収納ボックス、
VWR®, マイクロチューブ収納ボックス、
17 Options Available Below
Product Details & Documents
PP.
  • Highly transparent, with bold identifying colours and a distinctive wave design
  • Uniform wells with location ID; capacity indicated on the outside of the box
  • Black light-tight version available for light-sensitive samples
  • Frosted labelling patch
  • Alphanumeric identification on the bottom
Compact footprint for microtube and cryogenic vial storage. Store 1,5/2,0 ml microtubes or cryogenic vials. Complete with an easy open, friction-fit lid. Will fit into most standard freezer racking.
 : Assorted pack contains one of each colour blue, green, pink, yellow and orange.
Product Family Options
50-well boxes, hinged lid with snap-clasp
Product Information
  • Compartments
    Colour
    W×D×H ext.
    Availability
    Price
  • 50
    Assorted colours
    142×92×56 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 50
    Natural
    142×92×56 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
81-well boxes, friction fit lid
Product Information
  • Compartments
    Colour
    W×D×H ext.
    Availability
    Price
  • 81
    Assorted colours
    130×130×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 81
    Natural
    130×130×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 81
    Blue
    130×130×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 81
    Green
    130×130×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 81
    Pink
    130×130×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 81
    Yellow
    130×130×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 81
    Orange
    130×130×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 81
    Black
    130×130×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
100-well boxes, hinged lid with snap-clasp
Product Information
  • Compartments
    Colour
    W×D×H ext.
    Availability
    Price
  • 100
    Assorted colours
    141×151×56 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 100
    Natural
    141×151×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 100
    Blue
    141×151×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 100
    Green
    141×151×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 100
    Yellow
    141×151×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 100
    Orange
    141×151×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • 100
    Black
    141×151×47 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items