Avantor® ACE®, Capillary and Nano Column Coupler
Catalog # C0003
Supplier: Avantor
Specifications
- 説明:Capillary column coupler
- Pk:1
Specifications
About this item
Avantor® ACE® capillary/nano column couplers are suitable for connecting capillary and nano LC columns to stand-alone capillary/nano guard cartridges or for making similar LC connections.