Avantor® ACE®, Preparative Column Coupler
Catalog # C0002
Supplier: Avantor
Specifications
- 説明:Column coupler (for use with H0009)
- Pk:1
Specifications
About this item
Column coupler made from stainless steel/PEEK suitable for connecting preparative columns to stand-alone guard cartridge holder, or for making similar HPLC connections.
Ideally suited to connect 30 mm id preparative columns to H0009 cartridge holder.