Avantor® ACE® Phenyl-300, HPLC Columns, 10 µm
: Avantor
These ultra inert Avantor® ACE® Phenyl-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes available (3 µm, 5 µm and 10 µm) and complementary phases
- Designed for peptide and protein separations
- Excellent peak shape even at extremely low levels of additives such as TFA, making them ideal for LC/MS
- Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
- Catalog No:
- ACE-235-1010
- ACE-235-7530
- ACE-235-2510
- ACE-235-1510
- ACE-235-0510
- ACE-235-0520
- ACE-235-1020
- ACE-235-1220
- ACE-235-1520
- ACE-235-2520
- ACE-235-7520
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 5,3%
- 5,3%
- 5,3%
- 5,3%
- 5,3%
- 5,3%
- 5,3%
- 5,3%
- 5,3%
- 5,3%
- 5,3%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 400 bar
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Guard holders and accessories
Frequently Bought Together