Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm
: Avantor
Avantor® ACE® Excel® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
- Ultra-inert for maximum performance and reproducibility
- Ultra low bleed for improved LC/MS compatibility
- Wide range of particle sizes (1,7 to 10 µm) and complementary phases
- Method validation kit available containing 3 columns from different batches
Maximum recommended pH: 1,5 to 11,5
Max. pressure: 1000 bar
- Catalog No:
- EXL-1211-0202U
- EXL-1211-1003U
- EXL-1211-1002U
- EXL-1211-2046U
- EXL-1211-0346U
- EXL-1211-1246U
- EXL-1211-2503U
- EXL-1211-2502U
- EXL-1211-0503U
- EXL-1211-1046U
- EXL-1211-3546U
- EXL-1211-1546U
- EXL-1211-1503U
- EXL-1211-0302U
- EXL-1211-2546U
- EXL-1211-0546U
- EXL-1211-0502U
- EXL-1211-7546U
- EXL-1211-1502U
- EXL-1211-7503U
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- pH range:
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Frequently Bought Together