Avantor® ACE® SuperC18, HPLC Columns, 10 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
- Ultra-inert for maximum performance and reproducibility
- Ultra low bleed for improved LC/MS compatibility
- Wide range of particle sizes (1,7 to 10 µm) and complementary phases
Maximum recommended pH: 1,5 to 11,5
Max. pressure: 275 bar
Specifications
- Catalog No:
- ACE13110110GD
- ACE-1311-2546
- ACE-1311-1010
- ACE-1311-1510
- ACE-1311-2510
- ACE13110120GD
- ACE-1311-1520
- ACE-1311-0510
- ACE-1311-2530
- ACE-1311-2520
- ACE-1311-1020
- ACE-1311-1030
- ACE-1311-1220
- ACE-1311-1530
- ACE-1311-0520
- ACE-1311-0530
- ACE-1311-7520
- ACE-1311-7530
- ACE-1311-3030
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- pH range:
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)