Orders LinkContactUsLinkComponent
ビーカー, 使い捨て, PP
ビーカー, 使い捨て, PP
Supplier:   VWR Collection
ビーカー, 使い捨て, PP
Supplier:  VWR Collection
Certificates
ビーカー, 使い捨て, PP
ビーカー, 使い捨て, PP
3 Options Available Below
Product Details & Documents
PP, transparent.
  • Withstand temperatures up to 121 °C
  • Shatter-resistant
  • Easy-to-read graduations
  • Transparent for clear content visibility
Beakers feature a pouring spout for quick and easy dispensing. Highly resistant to strong acids. Stackable. Steam autoclavable.
注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • 容量
    目盛り範囲
    Availability
    Price
  • 15 ml
    5-15 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $35.42
    $35.42
    1 * 100 items
       $35.42
  • 30 ml
    5-30 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $41.26
    $41.26
    1 * 100 items
       $41.26
  • 50 ml
    5-50 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $46.13
    $46.13
    1 * 100 items
       $46.13