Orders LinkContactUsLinkComponent
Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 5 µm
Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 5 µm
   Sustainable Options Available
Supplier:   Avantor
Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 5 µm
Supplier:  Avantor
   Sustainable Options Available
Certificates
Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 5 µm
Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 5 µm
12 Options Available Below
Product Details & Documents
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (3 µm and 5 µm ) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems
A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.
Delivery information: Avantor® ACE® Bioanalytical Method Development Kits contain Avantor® ACE® C18-300, Avantor® ACE® C4-300 and Avantor® ACE® Phenyl-300 stainless steel columns.
Specifications for Product Family
Specifications
  • Catalog No:
  • MDKB-5-0502
  • MDKB-5-1002
  • MDKB-5-1502
  • MDKB-5-2502
  • MDKB-5-0503
  • MDKB-5-1003
  • MDKB-5-1503
  • MDKB-5-2503
  • MDKB-5-0546
  • MDKB-5-1046
  • MDKB-5-1546
  • MDKB-5-2546
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Max. pressure:
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • 275 bar
  • pH range:
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • 2,0 - 8,0
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Specifications
Product Family Options
Product Information
  • 説明
    カラム形式
    細孔サイズ
    粒子サイズ
    内径
    長さ
    Availability
    Price
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    2,10 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-0502
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    2,10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-1002
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    2,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-1502
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    2,10 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-2502
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    3,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-0503
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    3,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-1003
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    3,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-1503
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    3,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-2503
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    4,60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-0546
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-1046
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-1546
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Bioanalytical 300 Å MDK, C18-300, C4-300, Phenyl-300
    Method development kit
    300 Å
    5 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    MDKB-5-2546
    Environmentally Preferable
    Low Manufacturing Impact
    Max. pressure
    275 bar
    pH range
    2,0 - 8,0
    Environmental certification
    ISO 14001:2015 (530840 UM15)