Orders LinkContactUsLinkComponent
シューズカバー、PP/PE、アドバンスド
シューズカバー、PP/PE、アドバンスド
シューズカバー、PP/PE、アドバンスド
シューズカバー、PP/PE、アドバンスド
These shoe covers are manufactured from a specially formulated PP/PE microporous film. They provide excellent fluid and barrier protection.
  • Highly durable, long-lasting abrasion- and skid-resistance
  • Excellent particulate barrier
  • High coefficient of friction provides excellent traction
  • Non linting
  • Serged seams
  • Catalog No:
  • 414004-650
  • 414004-649
  • 414004-651
  • Garment application:
  • Laboratory
  • Laboratory
  • Laboratory
Product Family Options
  • Colour
    高さ
    長さ
  • Blue
    171 mm
    432 mm
    1 * 300 items
    1 * 300 items
    Specifications
    Cat. No.
    414004-650
    Garment application
    Laboratory
  • Blue
    159 mm
    394 mm
    1 * 300 items
    1 * 300 items
    Specifications
    Cat. No.
    414004-649
    Garment application
    Laboratory
  • Blue
    178 mm
    470 mm
    1 * 1 Each
    1 * 1 Each
    Specifications
    Cat. No.
    414004-651
    Garment application
    Laboratory