Orders LinkContactUsLinkComponent
ニトリルグローブ、VWR®製
ニトリルグローブ、VWR®製
Supplier:   VWR Collection
ニトリルグローブ、VWR®製
Supplier:  VWR Collection
Certificates
3 Options Available Below
Product Details & Documents
These ambidextrous examination gloves are ideal for use in industrial and laboratory applications as they protect against biohazards and a broad range of chemicals. They are made from a premium compounded nitrile for high manufacturing quality measures and product consistency.
  • Non sterile, powder-free, textured finish
  • Extra soft and comfortable for every day use
  • Rolled, beaded cuff allows easy donning
  • Protect hands from a broad range of chemicals
  • 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins
Specifications for Product Family
Specifications
  • Catalog No:
  • 82026-424
  • 82026-428
  • 82026-430
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured
  • Textured
  • Textured
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,1 mm
  • 0,1 mm
  • 0,1 mm
  • Finger thickness:
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • Palm thickness:
  • 0,12 mm
  • 0,12 mm
  • 0,12 mm
Specifications
Product Family Options
Product Information
  • 手袋のサイズ
    Colour
    長さ
    Packed
    Availability
    Price
  • S
    Blue
    229 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $14.61
    $14.61
    1 * 100 items
       $14.61
    Specifications:
    More
    Specifications
    Cat. No.
    82026-424
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    Cuff thickness
    0,1 mm
    Finger thickness
    0,14 mm
    Palm thickness
    0,12 mm
  • L
    Blue
    229 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $14.61
    $14.61
    1 * 100 items
       $14.61
    Specifications:
    More
    Specifications
    Cat. No.
    82026-428
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    Cuff thickness
    0,1 mm
    Finger thickness
    0,14 mm
    Palm thickness
    0,12 mm
  • XL
    Blue
    229 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $14.61
    $14.61
    1 * 100 items
       $14.61
    Specifications:
    More
    Specifications
    Cat. No.
    82026-430
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    Cuff thickness
    0,1 mm
    Finger thickness
    0,14 mm
    Palm thickness
    0,12 mm