Orders LinkContactUsLinkComponent
VWR®, 実験用ボトル、ワイドネック
VWR®, 実験用ボトル、ワイドネック
Supplier:  VWR Collection
Certificates
VWR®, 実験用ボトル、ワイドネック
VWR®, 実験用ボトル、ワイドネック
8 Options Available Below
Product Details & Documents
Bottles constructed of borosilicate 3.3 glass, in clear or amber.
  • Excellent chemical resistance, high temperature resistance
  • 80 mm opening allows easy cleaning
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The large opening can even accommodate large volume funnels, giving easy access to scoops or spatulas.
Amber to protect light-sensitive materials.
 : In compliance with DIN ISO 4796-1 standards.
 : Supplied with screw cap and pouring ring.
 : The bottles are packed in shrinked-wrapped modules.
Product Family Options
Clear glass
Product Information
  • 容量
    説明
    Thread
    W×D×H
    Colour
    Availability
    Price
  • 500 ml
    With PP screw cap
    GL 80
    94×94×139 mm
    Clear
    1 * 1 items
  • 1000 ml
    With PP screw cap
    GL 80
    105×105×185 mm
    Clear
    1 * 1 items
  • 2000 ml
    With PP screw cap
    GL 80
    115×115×258 mm
    Clear
    1 * 1 items
  • 5000 ml
    With PP screw cap
    GL 80
    160×160×357 mm
    Clear
    1 * 1 items
Amber glass
Product Information
  • 容量
    説明
    Thread
    W×D×H
    Colour
    Availability
    Price
  • 500 ml
    With LDPE screw cap
    GL 80
    94×94×139 mm
    Amber
    1 * 1 items
  • 1000 ml
    With LDPE screw cap
    GL 80
    105×105×185 mm
    Amber
    1 * 1 items
  • 2000 ml
    With LDPE screw cap
    GL 80
    115×115×258 mm
    Amber
    1 * 1 items
  • 5000 ml
    With LDPE screw cap
    GL 80
    160×160×357 mm
    Amber