VWR®, 細胞培養用インサートプレート、PC
About this item
PC, sterile, with lid. These insert plates feature a thin, semi-transparent polycarbonate membrane with pore sizes from 0,1 to 12,0 μm.
- Treated for optimal cell attachment
- Assembled with well plates
- Resistant to most fixing and staining agents
- Non pyrogenic
- Sterilised by electron beam irradiation
Temperature range: –20 to +50 °C.
Certifications: ISO 9001, ISO 13485.
Packaging: Individually packed in recyclable PET.
Caution: Inserts are not autoclavable.
19 Options Available Below
Product Details & Documents
PC, sterile, with lid. These insert plates feature a thin, semi-transparent polycarbonate membrane with pore sizes from 0,1 to 12,0 μm.
- Treated for optimal cell attachment
- Assembled with well plates
- Resistant to most fixing and staining agents
- Non pyrogenic
- Sterilised by electron beam irradiation
Temperature range: –20 to +50 °C.
Certifications: ISO 9001, ISO 13485.
Packaging: Individually packed in recyclable PET.
Caution: Inserts are not autoclavable.
Specifications for Product Family
Specifications
- Catalog No:
- 734-2715
- 734-2720
- 734-2716
- 734-2721
- 734-2722
- 734-2717
- 734-2726
- 734-2731
- 734-2727
- 734-2732
- 734-2733
- 734-2728
- 734-2737
- 734-2742
- 734-2738
- 734-2743
- 734-2744
- 734-2745
- 734-2739
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
- 説明Culture areaRecommended working volumeMembraneMembrane ØPacked細孔サイズAvailabilityPrice
- 6-well insert plate4,67 cm²2,6 mlTranslucent24 mm6/plate, 4 plates/case0,1 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2715Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTranslucent24 mm6/plate, 4 plates/case0,4 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2720Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTranslucent24 mm6/plate, 4 plates/case1,0 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2716Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTransparent24 mm6/plate, 4 plates/case3,0 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2721Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTransparent24 mm6/plate, 4 plates/case8,0 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2722Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTransparent24 mm6/plate, 4 plates/case12,0 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2717Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTranslucent12 mm12/plate, 4 plates/case0,1 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2726Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTranslucent12 mm12/plate, 4 plates/case0,4 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2731Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTranslucent12 mm12/plate, 4 plates/case1,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2727Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTransparent12 mm12/plate, 4 plates/case3,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2732Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTransparent12 mm12/plate, 4 plates/case8,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2733Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTransparent12 mm12/plate, 4 plates/case12,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2728Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTranslucent6,5 mm12/plate, 4 plates/case0,1 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2737Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTranslucent6,5 mm12/plate, 4 plates/case0,4 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2742Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTranslucent6,5 mm12/plate, 4 plates/case1,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2738Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTransparent6,5 mm12/plate, 4 plates/case3,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2743Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTransparent6,5 mm12/plate, 4 plates/case8,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2744Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTransparent6,5 mm12/plate, 4 plates/case5,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2745Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTransparent6,5 mm12/plate, 4 plates/case12,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2739Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact