VWR®, 細胞培養用インサートプレート、PET
Supplier: VWR Collection
About this item
Polyester, sterile, with lid. These insert plates feature a thin, microscopically transparent polyester membrane that is tissue culture-treated for optimal cell attachment and growth.
- Provides excellent cell visibility under phase-contrast microscopy
- Allows assessment of cell viability and monolayer formation
- Can be assembled with 6-, 12- and 24-well plates
- Available in five pore sizes from 0,1 to 8,0 µm
- Non pyrogenic
- Sterilised by electron beam irradiation
Temperature range: –20 to +50 °C.
: ISO 9001, ISO 13485.
: Individually packed in recyclable PET.
: Inserts are not autoclavable.
18 Options Available Below
Product Details & Documents
Polyester, sterile, with lid. These insert plates feature a thin, microscopically transparent polyester membrane that is tissue culture-treated for optimal cell attachment and growth.
- Provides excellent cell visibility under phase-contrast microscopy
- Allows assessment of cell viability and monolayer formation
- Can be assembled with 6-, 12- and 24-well plates
- Available in five pore sizes from 0,1 to 8,0 µm
- Non pyrogenic
- Sterilised by electron beam irradiation
Temperature range: –20 to +50 °C.
: ISO 9001, ISO 13485.
: Individually packed in recyclable PET.
: Inserts are not autoclavable.
Specifications for Product Family
Specifications
- Catalog No:
- 734-3404
- 734-2723
- 734-2719
- 734-2724
- 734-2725
- 734-3261
- 734-3405
- 734-2734
- 734-2730
- 734-2735
- 734-2736
- 734-3262
- 734-3406
- 734-3263
- 734-2746
- 734-2741
- 734-2747
- 734-2748
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
- 説明Culture areaRecommended working volumeMembraneMembrane ØPacked細孔サイズAvailabilityPrice
- 6-well insert plate4,67 cm²2,6 mlTranslucent24 mm4 plates, 6 inserts each0,1 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-3404Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTranslucent24 mm4 plates, 6 inserts each0,4 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2723Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTranslucent24 mm4 plates, 6 inserts each1,0 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2719Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTransparent24 mm4 plates, 6 inserts each3,0 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2724Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTransparent24 mm4 plates, 6 inserts each8,0 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2725Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well insert plate4,67 cm²2,6 mlTransparent24 mm4 plates, 6 inserts each0,4 µmLog in for availability1 * 24 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-3261Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTranslucent12 mm4 plates, 12 inserts each0,1 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-3405Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTranslucent12 mm4 plates, 12 inserts each0,4 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2734Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTranslucent12 mm4 plates, 12 inserts each1,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2730Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTransparent12 mm4 plates, 12 inserts each3,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2735Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTransparent12 mm4 plates, 12 inserts each8,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2736Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well insert plate1,12 cm²1,5 mlTransparent12 mm4 plates, 12 inserts each0,4 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-3262Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTranslucent6,5 mm4 plates, 12 inserts each0,1 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-3406Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTransparent6,5 mm4 plates, 12 inserts each0,4 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-3263Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTranslucent6,5 mm4 plates, 12 inserts each0,4 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2746Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTranslucent6,5 mm4 plates, 12 inserts each1,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2741Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTransparent6,5 mm4 plates, 12 inserts each3,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2747Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 24-well insert plate0,33 cm²0,6 mlTransparent6,5 mm4 plates, 12 inserts each8,0 µmLog in for availability1 * 48 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2748Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact