VWR®, 使い捨て紙製漏斗、
Disposable paper funnel, 186 ml
Catalog # 221-0274
Supplier: VWR Collection
Jump to:
Product Details & Documents
Made of clay-coated heavy paper stock.
- Innovative 'backrest' allows hands-free use when pouring viscous liquids
- Fits a wide variety of openings
- Maximum flexibility - Cut stem to any opening size or shape
- Apply a minimum of pressure and the funnel will stay in position
- Use with virtually any liquid or powder*
Good alternative to disposable plastic funnels. Environmentally friendly paper product will naturally biodegrade in the standard municipal waste stream. Helps to reduce cross contamination.
Caution: * Not recommended for hot liquids.
Specifications
- 容量:186 ml
- 説明:Disposable paper funnel
- Cat. No.:221-0274
- Environmentally Preferable:Sustainable Materials