Orders LinkContactUsLinkComponent
VWR®, ボトル、ウォーターサンプリング、
VWR®, ボトル、ウォーターサンプリング、
VWR®, ボトル、ウォーターサンプリング、
Supplier:  VWR Collection
Certificates
VWR®, ボトル、ウォーターサンプリング、
VWR®, ボトル、ウォーターサンプリング、
12 Options Available Below
Product Details & Documents
PET, virgin, food grade with PP caps.
  • Irradiated - sterile
  • Different coloured caps for easy indentification of test
  • Tamper-evident closure to ensure sampling integrity
  • Full traceability, batch number and individual bottle number on each label
  • Fully recyclable
  • Cap diameter: 32 mm
Lightweight, durable bottles. Tamper-evident screw caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Square bottles for better storage with limited space.
Blue cap: For natural, non chlorinated water without sodium thiosulphate.
Red cap: For chlorinated water dosed with 20 mg/L sodium thiosulphate.
Light blue cap: For chlorinated water dosed with 120 mg/L sodium thiosulphate.
Product Family Options
Product Information
  • 容量
    Dosage
    Cap colour
    W×D×H
    Packed
    Availability
    Price
  • 500 ml
    Undosed
    Blue
    70×70×192 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 126 items
    1 * 126 items
  • 500 ml
    Sodium thiosulphate (20 mg/L)
    Red
    70×70×192 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 126 items
    1 * 126 items
  • 500 ml
    Undosed
    Blue
    70×70×192 mm
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 108 items
    1 * 108 items
  • 500 ml
    Sodium thiosulphate (120 mg/L)
    Light blue
    70×70×192 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 126 items
    1 * 126 items
  • 500 ml
    Undosed
    Blue
    70×70×192 mm
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 126 items
    1 * 126 items
  • 500 ml
    Sodium thiosulphate (20 mg/L)
    Red
    70×70×192 mm
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 126 items
    1 * 126 items
  • 500 ml
    Sodium thiosulphate (20 mg/L)
    Red
    70×70×192 mm
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 112 items
    1 * 112 items
  • 500 ml
    Sodium thiosulphate (120 mg/L)
    Light blue
    70×70×192 mm
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 126 items
    1 * 126 items
  • 500 ml
    Sodium thiosulphate (20 mg/L)
    Light blue
    70×70×192 mm
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 112 items
    1 * 112 items
  • 1000 ml
    Undosed
    Blue
    89×89×222 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 84 items
    1 * 84 items
  • 1000 ml
    Sodium thiosulphate (20 mg/L)
    Red
    89×89×222 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 84 items
    1 * 84 items
  • 1000 ml
    Sodium thiosulphate (120 mg/L)
    Light blue
    89×89×222 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 84 items
    1 * 84 items