VWR® MediaBag TW Dehydrated Culture Media in Bags
Catalog # 7101.00450
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:100
- 説明:MediaBag TW for BPW
- Sterile water to be added:450 ml/bag
- For:450 ml
Specifications
About this item
MediaBags TW (Twirl-bag) are filled with pre-weighed granulated media, double-bagged bag and irradiated. Add sterile water under aseptic conditions.