VWR® Nitrile Extra Light, Nitrile Gloves
These thin, ambidextrous gloves are suitable for use in medical applications, food processing and for general lab requirements in non hazardous areas.
- Non sterile, powder-free, textured fingertips
- Very thin: Provide excellent elasticity for good wearer comfort
- Free from phthalates/softeners and allergenic latex proteins; particularly skin friendly for users with latex allergy
- Beaded cuffs for easier donning
- Low thickness and textured fingertips for excellent grip
: EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4; ASTM D6319, ASTM F1671; MDR Class 1; PPE Cat. III
Food contact approved.
Food contact approved.
: * Thickness is double measured, minimum values
- Catalog No:
- 112-4193
- 112-4194
- 112-4197
- 112-4196
- 112-4195
- Model:
- Nitrile Extra Light
- Nitrile Extra Light
- Nitrile Extra Light
- Nitrile Extra Light
- Nitrile Extra Light
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff thickness:
- 0,10 mm*
- 0,10 mm*
- 0,10 mm*
- 0,10 mm*
- 0,10 mm*
- Finger thickness:
- 0,14 mm*
- 0,14 mm*
- 0,14 mm*
- 0,14 mm*
- 0,14 mm*
- Palm thickness:
- 0,12 mm*
- 0,12 mm*
- 0,12 mm*
- 0,12 mm*
- 0,12 mm*
- EN norm:
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4
- EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4
- Food contact approved:
- +
- +
- +
- +
- +
Choosing the right glove for the job in hand Regulatory changes affecting our VWR collection gloves VWR Collection gloves pass thorough testing
Frequently Bought Together