VWR®, スパチュラ、
263 mm, White
Supplier: VWR Collection
Jump to:
Product Details & Documents
PS, white. This spatula, with its long, ergonomic solid handle and pointed blade, can easily pierce paper and plastic bags.
- Production, assembling and packaging according to cleanroom Class 7 (10000)
- According to EU food and FDA guidelines
- Individually packed, sterilised by gamma radiation
Ideal for sampling powder, granules and pastes.
Documents
Specifications
- Colour:White
- 長さ:263 mm