Orders LinkContactUsLinkComponent
VWR®, サンプリングスプーン
VWR®, サンプリングスプーン
Supplier:  VWR Collection
Certificates
VWR®, サンプリングスプーン
VWR®, サンプリングスプーン
3 Options Available Below
Product Details & Documents
PS, white. These disposable spoons are produced in a cleanroom environment and sterilised by gamma radiation. They are ideal for sampling powder, granules and pastes.
  • Available in two different sizes: 2,5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon)
  • Production, assembly and packaging according to cleanroom Class 7 (10000)
  • According to EU food and FDA guidelines
  • Individually packed, sterilised by gamma radiation
Product Family Options
Product Information
  • 長さ
    容量
    Availability
    Price
  • 127 mm
    2,5 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    231-0493
  • 170 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    231-0494
  • 170 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $141.80
    $141.80
    1 * 100 items
       $141.80
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU10819-505