VWR® ARISTAR®, ICP Multi-Element Solution diluted According USP 232 Oral dose, 8 Elements in 7% Nitric Acid
Catalog # 85037.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- パックタイプ:Plastic bottle
- Pk:100 mL
- 説明:ICP multi element solution diluted
Specifications
About this item
Eight elements in 7% nitric acid.
Cd 2,5 mg/L, Pb 5 mg/L, As 1,5 mg/L, Hg 1,5 mg/L, Mo 10 mg/L, Ni 50 mg/L, V 10 mg/L, Cu 100 mg/L in HNO₃ 7%.
Certifications: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Packaging: USP232 oral dose