VWR®, クリーンルーム用オーバーオール
Supplier: VWR Collection
About this item
These sterile, single-use overalls are made of breathable SMS fabric. They are ideal for use in cleanroom production and research.
- Hood and two-way closure
- Elasticated wrist, ankles and back for tighter fit and easier donning
- Thumb loops (latex-free)
- Bound seams
- Without boots
- Folded with snap release to arms and legs to aid aseptic donning
Suitable for use in ISO Class 5 + 6 cleanroom environments.
Weight: 60 g
Level I acc. IEST-RP-CC003.3
Certifications: Type 5/6; Cat. III; EN 1149-5.
Packaging: Individually double-packed in easy-tear bags. 20 overalls per liner.
13 Options Available Below
Product Details & Documents
These sterile, single-use overalls are made of breathable SMS fabric. They are ideal for use in cleanroom production and research.
- Hood and two-way closure
- Elasticated wrist, ankles and back for tighter fit and easier donning
- Thumb loops (latex-free)
- Bound seams
- Without boots
- Folded with snap release to arms and legs to aid aseptic donning
Suitable for use in ISO Class 5 + 6 cleanroom environments.
Weight: 60 g
Level I acc. IEST-RP-CC003.3
Certifications: Type 5/6; Cat. III; EN 1149-5.
Packaging: Individually double-packed in easy-tear bags. 20 overalls per liner.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 113-7881
- VWRA114-9324
- 113-7882
- VWRA114-9325
- 113-7883
- VWRA114-9326
- 113-7884
- VWRA114-9327
- 113-7885
- VWRA114-9328
- 113-7886
- VWRA114-9329
- VWRA114-9330
- Model:
- With hood
- With hood
- With hood
- With hood
- With hood
- With hood
- With hood
- With hood
- With hood
- With hood
- With hood
- With hood
- With hood
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Weight:
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
- 60 g
Specifications
Product Family Options
Product Information
- Colour衣服のサイズSterileAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-7881ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.VWRA114-9324ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.113-7882ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.VWRA114-9325ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.113-7883ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.VWRA114-9326ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.113-7884ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.VWRA114-9327ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.113-7885ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.VWRA114-9328ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.113-7886ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.VWRA114-9329ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g - Specifications:
More
Specifications
Cat. No.VWRA114-9330ModelWith hoodCleanroom class100/ISO 5Garment applicationCleanroom|LaboratoryWeight60 g