Orders LinkContactUsLinkComponent
VWR®, 遠心分離機用チューブ
VWR®, 遠心分離機用チューブ
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 遠心分離機用チューブ
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 遠心分離機用チューブ
3 Options Available Below
Product Details & Documents
PS, clear, conical, with flat cap with integral sealing.
  • Electron beam irradiated
  • Longer length screw caps with sealing ring prevent any leakage
  • Engraved graduations at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Specifications for Product Family
Specifications
  • Catalog No:
  • 525-0638
  • 525-0639
  • 525-0640
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • 容量
    Closure type
    RCF max.
    Version
    Packed
    Availability
    Price
  • 15 ml
    Screw cap
    3000×g
    non sterile
    Bulk, 500/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0638
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 15 ml
    Screw cap
    3000×g
    sterile
    25/rack, 20 packs/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0639
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 15 ml
    Screw cap
    3000×g
    sterile
    50/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0640
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact