Orders LinkContactUsLinkComponent
VWR®, 遠心分離機用チューブ、円錐底
VWR®, 遠心分離機用チューブ、円錐底
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 遠心分離機用チューブ、円錐底
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 遠心分離機用チューブ、円錐底
VWR®, 遠心分離機用チューブ、円錐底
9 Options Available Below
Product Details & Documents
PP, conical, with white HDPE flat cap or plug cap.
  • Certified non cytotoxic, nuclease-free and pyrogen-free
  • Caps available in two styles: Flat cap with a smooth surface for additional labelling options, or plug style cap for a secure seal when using shakers and horizontal incubators
  • Engraved graduation at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • Can be used for storing and freezing samples at temperatures down to –80 °C
  • Sustainable design based on virgin resins
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
These disposable conical bottom and free-standing tubes are extremely versatile, and can be used for routine tasks as well as high speed centrifuging. Rack-packed tubes are packaged in freezable, recyclable cardboard racks. Tubes are sterilised by irradiation. Tubes and caps are autoclavable and freezable.
Specifications for Product Family
Specifications
  • Catalog No:
  • 525-0604
  • 525-0605
  • 525-0606
  • 525-0607
  • 525-0610
  • 525-0612
  • 525-0608
  • 525-0609
  • 525-0611
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Conical bottom
Product Information
  • 容量
    Closure type
    Cap type
    RCF max.
    Version
    Packed
    Availability
    Price
  • 15 ml
    Screw cap
    Flat cap
    12500×g
    sterile
    50/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0604
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 15 ml
    Screw cap
    Plug style
    12500×g
    sterile
    50/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0605
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 15 ml
    Screw cap
    Flat cap
    12500×g
    sterile
    50/rack, 10 racks/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0606
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 15 ml
    Screw cap
    Plug style
    12500×g
    sterile
    50/rack, 10 racks/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0607
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 50 ml
    Screw cap
    Flat cap
    12500×g
    sterile
    50/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0610
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 50 ml
    Screw cap
    Plug style
    12500×g
    sterile
    50/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0612
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 50 ml
    Screw cap
    Flat cap
    12500×g
    sterile
    25/rack, 20 racks/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0608
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 50 ml
    Screw cap
    Plug style
    12500×g
    sterile
    25/rack, 20 racks/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0609
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
Freestanding
Product Information
  • 容量
    Closure type
    Cap type
    RCF max.
    Version
    Packed
    Availability
    Price
  • 50 ml
    Screw cap
    Flat cap
    12500×g
    sterile
    50/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0611
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M